accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50816-18-7

IUPAC Name: dec-9-en-1-yl acetate
Synonyms: 9-decenyl acetate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: PIQXMYAEJSMANF-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results