accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50816-20-1

IUPAC Name: 2-[(8-bromooctyl)oxy]oxane
Synonyms: 2-[(8-bromooctyl)oxy]oxane
Molecular Weight (g/mol): 293.25
Molecular Formula: C13H25BrO2
InChi Key: JCRBYQZIJFWGOO-UHFFFAOYNA-N
SMILES: BrCCCCCCCCOC1CCCCO1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)