accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50824-05-0

IUPAC Name: 1-(bromomethyl)-4-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: JDNPUJCKXLOHOW-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(CBr)C=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)