CAS RN 50834-78-1
IUPAC Name:
4-(4-methoxyphenyl)-2-methyl-1,3-thiazole
Synonyms:
4-(4-methoxyphenyl)-2-methyl-1,3-thiazole
Molecular Weight (g/mol):
205.28
Molecular Formula:
C11H11NOS
InChi Key:
OVKCVKLSCKKRJY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=CSC(C)=N1
1
–
1
of
1
results
1
4-(4-Methoxyphenyl)-2-methylthiazole, 97%, Thermo Scientific™
CAS: 50834-78-1 Molecular Formula: C11H11NOS Molecular Weight (g/mol): 205.275 MDL Number: MFCD00460502 InChI Key: OVKCVKLSCKKRJY-UHFFFAOYSA-N Synonym: 4-4-methoxyphenyl-2-methylthiazole,4-4-methoxy-phenyl-2-methyl-thiazole,4-4-methoxyphenyl-2-methyl-1,3-thiazole,thiazole, 4-4-methoxyphenyl-2-methyl,acmc-1ampl,2-methyl-4-4-methoxyphenyl thiazole,4-methoxy-1-2-methyl 1,3-thiazol-4-yl benzene PubChem CID: 768765 IUPAC Name: 4-(4-methoxyphenyl)-2-methyl-1,3-thiazole SMILES: CC1=NC(=CS1)C2=CC=C(C=C2)OC