accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHHO

CAS RN 5085-95-0

IUPAC Name: 3-[4-(3-hydroxyphenoxy)phenoxy]phenol
Synonyms: 3-[4-(3-hydroxyphenoxy)phenoxy]phenol
Molecular Weight (g/mol): 294.31
Molecular Formula: C18H14O4
InChi Key: CHBWEVDVYGBMEJ-UHFFFAOYSA-N
SMILES: OC1=CC(OC2=CC=C(OC3=CC=CC(O)=C3)C=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results