accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50877-44-6

IUPAC Name: 4-bromo-5-methyl-1-phenyl-1H-pyrazole
Synonyms: 4-bromo-5-methyl-1-phenylpyrazole
Molecular Weight (g/mol): 237.1
Molecular Formula: C10H9BrN2
InChi Key: GRERYMFLSYSEAY-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=NN1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results