accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 50890-67-0

IUPAC Name: 3,13-diazatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-one
Synonyms: 3,13-diazatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-one
Molecular Weight (g/mol): 182.18
Molecular Formula: C11H6N2O
InChi Key: PFMTUGNLBQSHQC-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CN=C2C2=NC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)