accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50893-36-2

IUPAC Name: 1-chloroethyl ethyl carbonate
Synonyms: 1-chloroethyl ethyl carbonate
Molecular Weight (g/mol): 152.57
Molecular Formula: C5H9ClO3
InChi Key: YVRGKFXJZCTTRB-UHFFFAOYNA-N
SMILES: CCOC(=O)OC(C)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results