accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHONH2HCl

CAS RN 50894-62-7

IUPAC Name: pyrrolidine-2-carboxamide hydrochloride
Synonyms: pyrrolidine-2-carboxamide hydrochloride
Molecular Weight (g/mol): 150.61
Molecular Formula: C5H11ClN2O
InChi Key: CSKSDAVTCKIENY-UHFFFAOYNA-N
SMILES: Cl.NC(=O)C1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)