accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 509072-57-5

IUPAC Name: (2,2-difluorocyclopropyl)methanol
Synonyms: (2,2-difluorocyclopropyl)methanol
Molecular Weight (g/mol): 108.09
Molecular Formula: C4H6F2O
InChi Key: XOLSMTBBIZDHSG-UHFFFAOYNA-N
SMILES: OCC1CC1(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results