accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50924-49-7

IUPAC Name: 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-hydroxy-1H-imidazole-4-carboxamide
Synonyms: mizoribine
Molecular Weight (g/mol): 259.22
Molecular Formula: C9H13N3O6
InChi Key: HZQDCMWJEBCWBR-UHFFFAOYNA-N
SMILES: NC(=O)C1=C(O)N(C=N1)C1OC(CO)C(O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results