CAS RN 5094-12-2
CAS RN 5094-12-2
IUPAC Name:
2-methyl-1H,2H,3H,4H,5H-pyrido[4,3-b]indole
Synonyms:
2-methyl-1H,3H,4H,5H-pyrido[4,3-b]indole
Molecular Weight (g/mol):
186.26
Molecular Formula:
C12H14N2
InChi Key:
FYHWPFXPFFPQRT-UHFFFAOYSA-N
SMILES:
CN1CCC2=C(C1)C1=CC=CC=C1N2
1
–
1
of
1
results
1
2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 98.0+%, TCI America™

CAS: 5094-12-2 Molecular Formula: C12H14N2 Molecular Weight (g/mol): 186.26 MDL Number: MFCD00460783 InChI Key: FYHWPFXPFFPQRT-UHFFFAOYSA-N PubChem CID: 97424 IUPAC Name: 2-methyl-1,3,4,5-tetrahydropyrido[4,3-b]indole SMILES: CN1CCC2=C(C1)C3=CC=CC=C3N2