accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOOOCH2

CAS RN 50940-49-3

IUPAC Name: 4-oxo-4-[2-(prop-2-enoyloxy)ethoxy]butanoic acid
Synonyms: hea succinate
Molecular Weight (g/mol): 216.19
Molecular Formula: C9H12O6
InChi Key: UZDMJPAQQFSMMV-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)OCCOC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results