accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50979-18-5

IUPAC Name: guanidine; sulfamic acid
Synonyms: guanidine; sulfamic acid
Molecular Weight (g/mol): 156.16
Molecular Formula: CH8N4O3S
InChi Key: LNEUSAPFBRDCPM-UHFFFAOYSA-N
SMILES: NC(N)=N.NS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results