accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5098-11-3

IUPAC Name: 4-amino-1H-imidazole-5-carbonitrile
Synonyms: 5-amino-3H-imidazole-4-carbonitrile
Molecular Weight (g/mol): 108.1
Molecular Formula: C4H4N4
InChi Key: XEPBRDBFOSKYCF-UHFFFAOYSA-N
SMILES: NC1=C(NC=N1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results