accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50996-85-5

IUPAC Name: 1,3-dihydroxy-2-(hydroxymethyl)propan-2-aminium decyl sulfate
Synonyms: htris decyl sulfate
Molecular Weight (g/mol): 359.48
Molecular Formula: C14H33NO7S
InChi Key: GQVPKTHOBIZUCH-UHFFFAOYSA-N
SMILES: [NH3+]C(CO)(CO)CO.CCCCCCCCCCOS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results