accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50998-17-9

IUPAC Name: 6-bromoquinoxaline
Synonyms: 6-bromoquinoxaline
Molecular Weight (g/mol): 209.05
Molecular Formula: C8H5BrN2
InChi Key: NOYFLUFQGFNMRB-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N=CC=NC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)