accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51-05-8

IUPAC Name: hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
Synonyms: procaine hydrochloride
Molecular Weight (g/mol): 272.77
Molecular Formula: C13H21ClN2O2
InChi Key: HCBIBCJNVBAKAB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)