CAS RN 51-56-9
IUPAC Name:
hydrogen (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-hydroxy-2-phenylacetate bromide
Synonyms:
hydrogen (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-hydroxy-2-phenylacetate bromide
Molecular Weight (g/mol):
356.26
Molecular Formula:
C16H22BrNO3
InChi Key:
DWSGTFTVBLXELC-MOTQWOLNSA-N
SMILES:
[H+].[Br-].CN1[C@H]2CC[C@@H]1CC(C2)OC(=O)C(O)C1=CC=CC=C1