CAS RN 51-64-9
CAS RN 51-64-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ S(+)-Amphetamine (dextro-Amphetamine) Solution, Sold by MilliporeSigma™ Supelco™
Suitable for use in urine drug testing, clinical toxicology, forensic analysis, and isomer identification by LC-MS/MS or GC/MS.