accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51-78-5

IUPAC Name: hydrogen 4-aminophenol chloride
Synonyms: P-aminophenol hcl
Molecular Weight (g/mol): 145.59
Molecular Formula: C6H8ClNO
InChi Key: RVGOBWDGAVAVPJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results