accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHClCl

CAS RN 510-15-6

IUPAC Name: ethyl 2,2-bis(4-chlorophenyl)-2-hydroxyacetate
Synonyms: Acar caswell no. 434 ethyl p,p'-dichlorobenzilate ethyldichlorobenzilate epa pesticide chemical code 028801 chlorbenzylate chlorobenzilate chlorbenzilat geigy 338 kop-mite
Molecular Weight (g/mol): 325.19
Molecular Formula: C16H14Cl2O3
InChi Key: RAPBNVDSDCTNRC-UHFFFAOYSA-N
SMILES: CCOC(=O)C(O)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results