accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSNHNH2N

CAS RN 51011-54-2

IUPAC Name: 6-chloro-2-hydrazinyl-1,3-benzothiazole
Synonyms: 6-chloro-2-hydrazinyl-1,3-benzothiazole
Molecular Weight (g/mol): 199.66
Molecular Formula: C7H6ClN3S
InChi Key: JBAIRAQEQLLZMN-UHFFFAOYSA-N
SMILES: NNC1=NC2=CC=C(Cl)C=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results