accessibility menu, dialog, popup

UserName

Viewing profile as user:
PBrBr

CAS RN 51044-13-4

IUPAC Name: [(4-bromophenyl)methyl]triphenylphosphanium bromide
Synonyms: [(4-bromophenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 512.23
Molecular Formula: C25H21Br2P
InChi Key: FQJYKXVQABPCRA-UHFFFAOYSA-M
SMILES: [Br-].BrC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)