accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNHN

CAS RN 51047-54-2

IUPAC Name: 1-(6-methoxypyridin-2-yl)piperazine
Synonyms: 1-(6-methoxypyridin-2-yl)piperazine
Molecular Weight (g/mol): 193.25
Molecular Formula: C10H15N3O
InChi Key: OILYGSQXSLUZSY-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=N1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results