accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOO

CAS RN 5105-78-2

IUPAC Name: 4-{[(benzyloxy)carbonyl]amino}butanoic acid
Synonyms: 4-{[(benzyloxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: STQMDRQJSNKUAW-UHFFFAOYSA-N
SMILES: OC(=O)CCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)