accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClHO

CAS RN 5106-98-9

IUPAC Name: 4-chloro-2-hydroxybenzoic acid
Synonyms: 4-chloro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: LWXFCZXRFBUOOR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)