accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51066-74-1

IUPAC Name: dimethylazanium iodide
Synonyms: dimethylazanium iodide
Molecular Weight (g/mol): 173
Molecular Formula: C2H8IN
InChi Key: JMXLWMIFDJCGBV-UHFFFAOYSA-N
SMILES: [I-].C[NH2+]C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)