accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51085-91-7

IUPAC Name: 6-bromo-4-oxo-4H-chromene-3-carboxylic acid
Synonyms: 6-bromo-4-oxochromene-3-carboxylic acid
Molecular Weight (g/mol): 269.05
Molecular Formula: C10H5BrO4
InChi Key: IJTWNMVKNJSGMS-UHFFFAOYSA-N
SMILES: OC(=O)C1=COC2=CC=C(Br)C=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results