accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 5111-70-6

IUPAC Name: 5-methoxy-2,3-dihydro-1H-inden-1-one
Synonyms: 5-methoxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: QOPRWBRNMPANKN-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(=O)CCC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)