accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOCH3CH3CH3CH3H3C

CAS RN 51115-67-4

IUPAC Name: N,2,3-trimethyl-2-(propan-2-yl)butanamide
Synonyms: trimethyl isopropyl butanamide methyl diisopropyl propionamide
Molecular Weight (g/mol): 171.28
Molecular Formula: C10H21NO
InChi Key: RWAXQWRDVUOOGG-UHFFFAOYSA-N
SMILES: CNC(=O)C(C)(C(C)C)C(C)C
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results