accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOSOOOSOHOHOHOOHK(E/Z)

CAS RN 5115-71-9

IUPAC Name: potassium (2-phenyl-1-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanyl}ethylidene)amino sulfate
Synonyms: potassium (2-phenyl-1-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanyl}ethylidene)amino sulfate
Molecular Weight (g/mol): 447.51
Molecular Formula: C14H18KNO9S2
InChi Key: UYCWNAZWHVREMO-UHFFFAOYNA-M
SMILES: [K+].OCC1OC(SC(CC2=CC=CC=C2)=NOS([O-])(=O)=O)C(O)C(O)C1O
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)