accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51163-29-2

CH3NCOCH3

CAS RN 51163-29-2

IUPAC Name: 1-isocyanato-2,4-dimethylbenzene
Synonyms: 1-isocyanato-2,4-dimethylbenzene
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: QUOBVYPFBJUOAJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results