accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOOOOOOH

CAS RN 5117-19-1

IUPAC Name: 3,6,9,12,15,18,21-heptaoxatricosane-1,23-diol
Synonyms: octaethylene glycol ped-diol (n=8)
Molecular Weight (g/mol): 370.44
Molecular Formula: C16H34O9
InChi Key: GLZWNFNQMJAZGY-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCOCCOCCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results