accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51173-05-8

IUPAC Name: 5-fluoro-1,2-dihydropyridin-2-one
Synonyms: 5-fluoro-1H-pyridin-2-one
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: KLULSYPVWLJZAO-UHFFFAOYSA-N
SMILES: FC1=CNC(=O)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results