accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 512-42-5

IUPAC Name: sodium methyl sulfate
Synonyms: methyl bisulfate
Molecular Weight (g/mol): 134.08
Molecular Formula: CH3NaO4S
InChi Key: DZXBHDRHRFLQCJ-UHFFFAOYSA-M
SMILES: [Na+].COS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)