accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOH3COH3C

CAS RN 512-56-1

IUPAC Name: trimethyl phosphate
Synonyms: trimethyl phosphate|trimethyl-phosphate|trimethylphosphate|TMPA|phosphate, trimethyl-|TMPO|trimethyl orthophosphate|trimethylfosfat|ccris 610
Molecular Weight (g/mol): 140.08
Molecular Formula: C3H9O4P
InChi Key: WVLBCYQITXONBZ-UHFFFAOYSA-N
SMILES: COP(=O)(OC)OC
Molecular Weight (g/mol) 
  • (1)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)