accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSiOSiOSi

CAS RN 512-63-0

IUPAC Name: hexaphenyl-1,3,5,2,4,6-trioxatrisilinane
Synonyms: hexaphenyl-1,3,5,2,4,6-trioxatrisilinane
Molecular Weight (g/mol): 594.89
Molecular Formula: C36H30O3Si3
InChi Key: VCYDUTCMKSROID-UHFFFAOYSA-N
SMILES: O1[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)