accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51207-98-8

IUPAC Name: N-butyl-4-nitrobenzamide
Synonyms: N-butyl-4-nitrobenzamide
Molecular Weight (g/mol): 222.24
Molecular Formula: C11H14N2O3
InChi Key: ZPYLRHGQFFDNOH-UHFFFAOYSA-N
SMILES: CCCCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results