accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51227-30-6

IUPAC Name: 1-(4-methylpyridin-3-yl)ethan-1-one
Synonyms: 1-(4-methylpyridin-3-yl)ethanone
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: LCEINQHGZAFADC-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results