accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51294-75-8

IUPAC Name: 4-bromo-3,5-dimethyl-1-phenyl-1H-pyrazole
Synonyms: 4-bromo-3,5-dimethyl-1-phenylpyrazole
Molecular Weight (g/mol): 251.13
Molecular Formula: C11H11BrN2
InChi Key: GNXWITGSOFQXDG-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1Br)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results