accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClCl

CAS RN 513-88-2

IUPAC Name: 1,1-dichloropropan-2-one
Synonyms: 1,1-dichloropropanone
Molecular Weight (g/mol): 126.96
Molecular Formula: C3H4Cl2O
InChi Key: CSVFWMMPUJDVKH-UHFFFAOYSA-N
SMILES: CC(=O)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results