accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClH2N

CAS RN 5131-60-2

IUPAC Name: 4-chlorobenzene-1,3-diamine
Synonyms: 1,3-benzenediamine, 4-chloro-
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: ZWUBBMDHSZDNTA-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)