accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51310-54-4

IUPAC Name: 2-(trifluoromethyl)-1H-indole
Synonyms: 2-(trifluoromethyl)-1H-indole
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: QFHVHZJGQWMBTE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)