CAS RN 51350-19-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ EHNA, Hydrochloride
The racemic mixture of a cell-permeable adenine derivative that acts as a PDE II-selective phosphodiesterase inhibitor and adenosine deaminase.