accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2N

CAS RN 51387-90-7

IUPAC Name: 2-(1-methylpyrrolidin-2-yl)ethan-1-amine
Synonyms: 2-(1-methylpyrrolidin-2-yl)ethanamine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: PNHGJPJOMCXSKN-UHFFFAOYNA-N
SMILES: CN1CCCC1CCN
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results