CAS RN 514-36-3
IUPAC Name:
2-[(1R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonyms:
fludrocortisone acetate florinef acetate cortineff fluorocortisone acetate fludrocortisone 21-acetate alflorone acetate cortef-F scherofluron fluohydrocortisone acetate fluohydrisone acetate preparation
Molecular Weight (g/mol):
422.49
Molecular Formula:
C23H31FO6
InChi Key:
SYWHXTATXSMDSB-GSLJADNHSA-N
SMILES:
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
1
–
1
of
1
results
1
Thermo Scientific Chemicals Fludrocortisone acetate
CAS: 514-36-3 Molecular Formula: C23H31FO6 Molecular Weight (g/mol): 422.49 InChI Key: SYWHXTATXSMDSB-GSLJADNHSA-N PubChem CID: 124040734