accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51417-13-1

IUPAC Name: 5-bromo-1,3-dimethyl-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-1,3-dimethylpyridin-2-one
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: UEHHXNZKBMALIY-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C=C(C)C1=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)