accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5142-22-3

IUPAC Name: 1-methyl-1H-purin-6-amine
Synonyms: 1-methyl-1H-purin-6-amine
Molecular Weight (g/mol): 149.16
Molecular Formula: C6H7N5
InChi Key: HPZMWTNATZPBIH-UHFFFAOYSA-N
SMILES: CN1C=NC2=NC=NC2=C1N
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)