CAS RN 5144-89-8
1,10-Phenanthroline, 0.1% (w/v), Ricca Chemical
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline monohydrate, 99+%
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N PubChem CID: 21226 SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline monohydrate, ACS
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline;hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline monohydrate, 99+%, pure
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N PubChem CID: 21226 SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline monohydrate, ACS reagent
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N PubChem CID: 21226 SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline monohydrate, ACS, MilliporeSigma™
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
MilliporeSigma™ Calbiochem™ o-Phenanthroline
A metal chelating agent that prevents the induction of chromosomal aberrations in streptozotocin-treated cells.
1,10-Phenanthroline, 0.25% (w/v), Ricca Chemical
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline, 0.3% (w/v), Ricca Chemical
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline, 0.1% (w/v) Alcoholic, Ricca Chemical
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline Monohydrate 99.0+%, TCI America™
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline Monohydrate, ACS Reagent Grade, Ricca Chemical
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline Monohydrate 99.0+%, TCI America™
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1,10-Phenanthroline Monohydrate 98.0+%, TCI America™
CAS: 5144-89-8 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00149973 InChI Key: PPQJCISYYXZCAE-UHFFFAOYSA-N Synonym: 1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate PubChem CID: 21226 IUPAC Name: 1,10-phenanthroline hydrate SMILES: O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
- 1
- 2